Vidarabine monohydrate 98%

Supplier: Ambeed
Danger

Synonyms: (2R,3S,4S,5R)-2-(6-Amino-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol hydrate, Ara-A, Adenine 9-beta-D-Arabinofuranoside, 9-(beta-D-Arabinofuranosyl)adenine

A586864-1MG A586864-25MG A586864-5MG
76790-446EA 5.31 USD
76790-446 76790-448 76790-450
Vidarabine monohydrate 98%
Vidarabine monohydrate

Precaución: for Research Use Only

Formula: C₁₀H₁₃N₅O₄·H₂O
MW: 285.26 g/mol
MDL Number: MFCD00150980
CAS Number: 24356-66-9
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR