4-Chloro-2-methylthieno[3,2-d]pyrimidine 95%

Supplier: Ambeed
Warning

A586802-1G A586802-250MG A586802-5G
76790-440EA 113.52 USD
76790-440 76790-442 76790-444
4-Chloro-2-methylthieno[3,2-d]pyrimidine 95%
4-Chloro-2-methylthieno[3,2-d]pyrimidine

Precaución: for Research Use Only

Formula: C₇H₅ClN₂S
MW: 184.646 g/mol
MDL Number: MFCD09834968
CAS Number: 319442-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR