Tricyclo[5.2.1.0(2,6)]-8-decanone 98%

Supplier: Ambeed
Warning

Synonyms: Hexahydro-1H-4,7-methanoinden-5(6H)-one, Tricyclo[5.2.1.0(2,6)]decan-8-one

A583020-5G A583020-25G
76790-428EA 12.74 USD
76790-428 77669-534
Tricyclo[5.2.1.0(2,6)]-8-decanone 98%
Tricyclo[5.2.1.0(2,6)]-8-decanone

Precaución: for Research Use Only

Formula: C₁₀H₁₄O
MW: 150.22 g/mol
MDL Number: MFCD00168034
CAS Number: 13380-94-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR