Trimellitic anhydride chloride 98%

Supplier: Ambeed
Danger

Synonyms: 1,3-Dioxo-1,3-dihydroisobenzofuran-5-carbonyl chloride, 1,2,4-Benzenetricarboxylic acid 1,2-anhydride 4-chloride

A580265-25G A580265-5G
76790-424EA 13.8 USD
76790-424 76790-426
Trimellitic anhydride chloride 98%
Trimellitic anhydride chloride

Precaución: for Research Use Only

Formula: C₉H₃ClO₄
MW: 210.57 g/mol
MDL Number: MFCD00005924
CAS Number: 1204-28-0
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR