Sudan III 96%

Supplier: Ambeed
Warning

Synonyms: Solvent Red 23, Sudan® III (Red), Scarlet B, Sudan Red III, Cerasin Red, Sudan Red BK, Sudan Red B, Fat Ponceau G, Sudan G, D and C Red No. 17, Oil red, Tony Red, Oil scarlet, Organol® red BS, 1-(4-(Phenylazo)phenylazo)-2-naphthol, Sudan V, 1-((4-(Phenyldiazenyl)phenyl)diazenyl)naphthalen-2-ol

A579825-1MG A579825-25G A579825-100G A579825-500G
76790-410EA 6 USD
76790-410 76790-412 77669-522 77872-959
Sudan III 96%
Sudan III

Precaución: for Research Use Only

Formula: C₂₂H₁₆N₄O
MDL Number: MFCD00003905
CAS Number: 85-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR