1-Boc-pyrrole-2-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-formyl-1H-pyrrole-1-carboxylate

A579268-250MG A579268-5G
76790-406EA 23.34 USD
76790-406 76790-408
1-Boc-pyrrole-2-carboxaldehyde 95%
1-Boc-pyrrole-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₃
MW: 195.22
MDL Number: MFCD03086220
CAS Number: 161282-57-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR