Sulfolane 97%

Supplier: Ambeed
Warning

A576973-100G A576973-25G A576973-500G
76790-384EA 10 USD
76790-384 76790-386 76790-388
Sulfolane 97%
Sulpholane

Precaución: for Research Use Only

Formula: C₄H₈O₂S
MDL Number: MFCD00005484
CAS Number: 126-33-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR