(R)-(2'-Isopropoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine 98%

Supplier: Ambeed
Warning

A576871-100MG A576871-1G A576871-250MG
76790-378EA 106.09 USD
76790-378 76790-380 76790-382
(R)-(2'-Isopropoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine 98%
(R)-(2'-Isopropoxy-[1,1'-binaphthalen]-2-yl)diphenylphosphine

Precaución: for Research Use Only

Formula: C₃₅H₂₉OP
MW: 496.58 g/mol
MDL Number: MFCD11111447
CAS Number: 189274-36-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR