Triphenylphosphine sulphide 99%

Supplier: Ambeed
Warning

A576741-1000G A576741-100G A576741-25G A576741-500G A576741-5G A576741-10G
76790-368EA 472 USD
76790-368 76790-370 76790-372 76790-374 76790-376 77602-426
Triphenylphosphine sulphide 99%
Triphenylphosphine sulphide

Precaución: for Research Use Only

Formula: C₁₈H₁₅PS
MW: 294.36 g/mol
MDL Number: MFCD00004917
CAS Number: 3878-45-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR