Flavanone 98%

Supplier: Ambeed
Warning

Synonyms: 2-Phenylchroman-4-one

A570598-100MG A570598-10MG A570598-25MG A570598-50MG
76790-340EA 11.67 USD
76790-340 76790-342 76790-344 76790-346
Flavanone 98%
Flavanone

Precaución: for Research Use Only

Formula: C₁₅H₁₂O₂
MW: 224.26
MDL Number: MFCD00006841
CAS Number: 487-26-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR