(4-Cyanophenyl)acetic acid ethyl ester 98%

Supplier: Ambeed
Danger

Synonyms: Ethyl (4-cyanophenyl)acetate, Ethyl 2-(4-cyanophenyl)acetate

A567686-1G A567686-25G A567686-5G A567686-10G
76790-322EA 12 USD
76790-322 76790-324 76790-326 77872-948
(4-Cyanophenyl)acetic acid ethyl ester 98%
(4-Cyanophenyl)acetic acid ethyl ester

Precaución: for Research Use Only

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD09953133
CAS Number: 1528-41-2
UN: 3439
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR