[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
(2-(1,3-Dioxolan-2-yl)ethyl)triphenylphosphonium bromide
A566804-100G
A566804-25G
A566804-5G
76790-316EA
224.92
USD
76790-316
76790-318
76790-320
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide 98%
[2-(1,3-Dioxolan-2-yl)ethyl]triphenylphosphonium bromide
Precaución: for Research Use Only
Formula:
C₂₃H₂₄BrO₂P MW: 443.3132 g/mol |
MDL Number:
MFCD00075119 CAS Number: 86608-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...