Tetrafluorobenzene-1,2-diol 97%

Supplier: Ambeed
Warning

A560703-1G A560703-250MG A560703-5G
76790-302EA 318 USD
76790-302 76790-304 77602-386
Tetrafluorobenzene-1,2-diol 97%
Tetrafluorobenzene-1,2-diol

Precaución: for Research Use Only

Formula: C₆H₂F₄O₂
MDL Number: MFCD01075279
CAS Number: 1996-23-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR