1-(Dimethylamino)-2-methylpentan-3-one 95%

Supplier: Ambeed
Warning

A559994-1G A559994-25G
76790-298EA 5.31 USD
76790-298 76790-300
1-(Dimethylamino)-2-methylpentan-3-one 95%
1-(Dimethylamino)-2-methylpentan-3-one

Precaución: for Research Use Only

Formula: C₈H₁₇NO
MW: 143.2267 g/mol
MDL Number: MFCD13185946
CAS Number: 51690-03-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR