4-n-Pentyloxyphenyl-trans-4-ethylcyclohexanecarboxylate 95+%

Supplier: Ambeed
Warning

Synonyms: Trans-4-(pentyloxy)phenyl 4-ethylcyclohexanecarboxylate

A558699-100G A558699-25G A558699-5G
76790-288EA 179.3 USD
76790-288 76790-290 76790-292
4-n-Pentyloxyphenyl-trans-4-ethylcyclohexanecarboxylate 95+%
4-n-Pentyloxyphenyl-trans-4-ethylcyclohexanecarboxylate

Precaución: for Research Use Only

Formula: C₂₀H₃₀O₃
MW: 318.457 g/mol
MDL Number: MFCD22125200
CAS Number: 67589-53-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR