Methyl-N-acetyl-L-tryptophanate 98%

Supplier: Ambeed
Warning

Synonyms: N-Acetyl-L-tryptophan methyl ester, Methyl N-acetyl-L-tryptophanate, Ac-Trp-OMe

A555995-5G A555995-1G A555995-100G A555995-25G
76790-280EA 43.5 USD
76790-280 77602-364 77669-466 77669-468
Methyl-N-acetyl-L-tryptophanate 98%
Methyl-N-acetyl-L-tryptophanate

Precaución: for Research Use Only

Formula: C₁₄H₁₆N₂O₃
MW: 260.293 g/mol
MDL Number: MFCD00022759
CAS Number: 2824-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR