2-Amino-3-pyridineboronic acid pinacol ester 95%

Supplier: Ambeed
Danger

Synonyms: 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-amine

A555211-1G A555211-250MG A555211-100MG A555211-25G
76790-276EA 43 USD
76790-276 76790-278 77872-929 77872-930
2-Amino-3-pyridineboronic acid pinacol ester 95%
2-Amino-3-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₇BN₂O₂
MDL Number: MFCD09260488
CAS Number: 1073354-97-8
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR