Canertinib 97%

Supplier: Ambeed
Warning

Synonyms: N-(4-((3-Chloro-4-fluorophenyl)amino)-7-(3-morpholinopropoxy)quinazolin-6-yl)acrylamide

A554853-10MG A554853-250MG A554853-100MG A554853-1G A554853-50MG
76790-274EA 65 USD
76790-274 77602-360 77669-454 77669-456 77669-458
Canertinib 97%
Canertinib

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClFN₅O₃
MW: 486 g/mol
MDL Number: MFCD09837878
CAS Number: 267243-28-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR