Triethyl 1,3,5-triazine-2,4,6-tricarboxylate 98%

Supplier: Ambeed
Warning

A554395-1G A554395-250MG
76790-270EA 139 USD
76790-270 76790-272
Triethyl 1,3,5-triazine-2,4,6-tricarboxylate 98%
Triethyl 1,3,5-triazine-2,4,6-tricarboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₅N₃O₆
MW: 297.26 g/mol
MDL Number: MFCD01419268
CAS Number: 898-22-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR