2-(4-Chloro-2-nitrophenyl)acetic acid 98%

Supplier: Ambeed
Warning

A553665-100G A553665-5G
76790-266EA 277 USD
76790-266 77669-444
2-(4-Chloro-2-nitrophenyl)acetic acid 98%
2-(4-Chloro-2-nitrophenyl)acetic acid

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄
MDL Number: MFCD11036386
CAS Number: 37777-71-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR