2,4,6-Triphenylboroxin 98%

Supplier: Ambeed
Warning

A552594-100G A552594-1G A552594-25G A552594-5G
76790-258EA 109 USD
76790-258 76790-260 76790-262 76790-264
2,4,6-Triphenylboroxin 98%
2,4,6-Triphenylboroxin

Precaución: for Research Use Only

Formula: C₁₈H₁₅B₃O₃
MDL Number: MFCD00014587
CAS Number: 3262-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR