2-Methoxy-5-(trifluoromethoxy)benzyl alcohol 95%

Supplier: Ambeed
Warning

A544933-1G A544933-250MG A544933-5G
76790-234EA 38 USD
76790-234 76790-236 76790-238
2-Methoxy-5-(trifluoromethoxy)benzyl alcohol 95%
2-Methoxy-5-(trifluoromethoxy)benzyl alcohol

Precaución: for Research Use Only

Formula: C₉H₉F₃O₃
MDL Number: MFCD04115960
CAS Number: 562840-50-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR