(S)-Methyl 1-methyl-5-oxopyrrolidine-2-carboxylate 95%

Supplier: Ambeed
Warning

A541115-100MG A541115-1G A541115-250MG
76790-222EA 7 USD
76790-222 76790-224 76790-226
(S)-Methyl 1-methyl-5-oxopyrrolidine-2-carboxylate 95%
(S)-Methyl 1-methyl-5-oxopyrrolidine-2-carboxylate

Precaución: for Research Use Only

Formula: C₇H₁₁NO₃
MW: 157.16 g/mol
MDL Number: MFCD12026427
CAS Number: 42435-88-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR