N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine 95%

Supplier: Ambeed
Warning

A539105-100MG A539105-1MG A539105-250MG A539105-50MG A539105-5G A539105-1G
76790-212EA 78 USD
76790-212 76790-214 76790-216 76790-218 76790-220 77872-904
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine 95%
N,N,N',N'-Tetrakis(2-pyridylmethyl)ethylenediamine

Precaución: for Research Use Only

Formula: C₂₆H₂₈N₆
MDL Number: MFCD00036918
CAS Number: 16858-02-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR