Trimellitic acid tris(2-ethylhexyl) ester 98%
Supplier: Ambeed
|
Warning
|
A538685-1000G
A538685-100G
A538685-500G
76790-206EA
25
USD
76790-206
76790-208
76790-210
Trimellitic acid tris(2-ethylhexyl) ester 98%
Trimellitic acid tris(2-ethylhexyl) ester
Precaución: for Research Use Only
Formula:
C₃₃H₅₄O₆ |
MDL Number:
MFCD00053799 CAS Number: 3319-31-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...