1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS 98%

Supplier: Ambeed
Warning

Synonyms: CTC-Q-PHOS, 1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene, QPhos, 1,2,3,4,5-Pentaphenyl-1'-(di-t-butylphosphino)ferrocene

A536604-100MG A536604-1G A536604-250MG A536604-50MG A536604-5G A536604-25G
76790-192EA 15 USD
76790-192 76790-194 76790-196 76790-198 76790-200 77872-897
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS 98%
1,2,3,4,5-Pentaphenyl-1'-(di-tert-butylphosphino)ferrocene CTC-Q-PHOS

Precaución: for Research Use Only

Formula: C₄₈H₄₇FeP
MW: 710.706 g/mol
MDL Number: MFCD03788940
CAS Number: 312959-24-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR