(5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-((2'-(1-trityl-1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-imidazole-5-carboxylate 95%
Supplier: Ambeed
|
Danger
|
Synonyms:
Tritylolmesartan Medoxomil
A536233-10G
A536233-25G
A536233-5G
76790-186EA
43.5
USD
76790-186
76790-188
76790-190
(5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-((2'-(1-trityl-1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-imidazole-5-carboxylate 95%
(5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 4-(2-hydroxypropan-2-yl)-2-propyl-1-((2'-(1-trityl-1H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1H-imidazole-5-carboxylate
Precaución: for Research Use Only
Formula:
C₄₈H₄₄N₆O₆ MW: 800.89 g/mol |
MDL Number:
MFCD08704231 CAS Number: 144690-92-6 UN: 1325 ADR: 4.1,Ⅲ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...