2-(Dimethylamino)benzaldehyde 98%

Supplier: Ambeed
Warning

A536030-250MG A536030-5G
76790-184EA 22.28 USD
76790-184 77572-696
2-(Dimethylamino)benzaldehyde 98%
2-(Dimethylamino)benzaldehyde

Precaución: for Research Use Only

Formula: C₉H₁₁NO
MW: 149.19 g/mol
MDL Number: MFCD03407377
CAS Number: 579-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR