2-Butyl-3-((2'-(2-trityl-2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1,3-diazaspiro[4.4]non-1-en-4-one 97%

Supplier: Ambeed
Danger

Synonyms: Irbesartan N-Trityl Impurity

A532493-100MG A532493-1MG A532493-1G
76790-170EA 9 USD
76790-170 76790-172 77669-364
2-Butyl-3-((2'-(2-trityl-2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1,3-diazaspiro[4.4]non-1-en-4-one 97%
2-Butyl-3-((2'-(2-trityl-2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-1,3-diazaspiro[4.4]non-1-en-4-one

Precaución: for Research Use Only

Formula: C₄₄H₄₂N₆O
CAS Number: 886999-35-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR