Methyl 2,5-dimethyl-3-furancarboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Methyl 2,5-dimethylfuran-3-carboxylate

A531376-1G A531376-25G A531376-5G
76790-154EA 22.28 USD
76790-154 76790-156 77669-360
Methyl 2,5-dimethyl-3-furancarboxylate 95%
Methyl 2,5-dimethyl-3-furancarboxylate

Precaución: for Research Use Only

Formula: C₈H₁₀O₃
MW: 154.163 g/mol
MDL Number: MFCD00010303
CAS Number: 6148-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR