Cyclobutane-1,1-dicarboxylic acid ethyl ester 98%

Supplier: Ambeed
Warning

Synonyms: 1-(Ethoxycarbonyl)cyclobutanecarboxylic acid

A529941-1G A529941-5G
76790-146EA 30.77 USD
76790-146 76790-148
Cyclobutane-1,1-dicarboxylic acid ethyl ester 98%
Cyclobutane-1,1-dicarboxylic acid ethyl ester

Precaución: for Research Use Only

Formula: C₈H₁₂O₄
MW: 172.18 g/mol
MDL Number: MFCD10699115
CAS Number: 54450-84-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR