1-Methyl-1H-indazol-3-amine 96%

Supplier: Ambeed

Synonyms: 3-Amino-1-methylindazole, 1-methyl-1H-indazole-3-ylamine, 3-Amino-1-methyl-1H-indazole

A528215-1G A528215-250MG A528215-5G
76790-140EA 24 USD
76790-140 76790-142 76790-144
1-Methyl-1H-indazol-3-amine 96%
1-Methyl-1H-indazol-3-amine

Precaución: for Research Use Only

Formula: C₈H₉N₃
MDL Number: MFCD09054755
CAS Number: 60301-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR