(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine 97%

Supplier: Ambeed
Warning

Synonyms: (1S,2S)-N1,N2-Bis(3,3-dimethylbutyl)-N1,N2-dimethylcyclohexane-1,2-diamine

A527751-250MG A527751-1G A527751-5G
76790-138EA 13 USD
76790-138 77602-252 77669-338
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine 97%
(1S,2S)-N,N'-Dimethyl-N,N'-bis(3,3-dimethylbutyl)cyclohexane-1,2-diamine

Precaución: for Research Use Only

Formula: C₂₀H₄₂N₂
MW: 310.57 g/mol
MDL Number: MFCD11869717
CAS Number: 767291-67-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR