2'-Methylbiphenyl-4-carbaldehyde 98%

Supplier: Ambeed
Danger

Synonyms: 2'-Methyl-[1,1'-biphenyl]-4-carbaldehyde

A527475-1G A527475-250MG A527475-5G
76790-134EA 60.48 USD
76790-134 76790-136 77602-250
2'-Methylbiphenyl-4-carbaldehyde 98%
2'-Methylbiphenyl-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₁₄H₁₂O
MW: 196.25 g/mol
MDL Number: MFCD01631915
CAS Number: 108934-21-0
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR