2-(Boc-amino)pyridine-5-carboxaldehyde 95%
Supplier: Ambeed
|
Warning
|
A527165-100MG
A527165-1G
A527165-250MG
A527165-5G
76790-126EA
19
USD
76790-126
76790-128
76790-130
76790-132
2-(Boc-amino)pyridine-5-carboxaldehyde 95%
2-(Boc-amino)pyridine-5-carboxaldehyde
Precaución: for Research Use Only
Formula:
C₁₁H₁₄N₂O₃ |
MDL Number:
MFCD08064228 CAS Number: 199296-40-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...