3-(Trifluoromethyl)benzhydrol 97%

Supplier: Ambeed
Warning

Synonyms: Phenyl(3-(trifluoromethyl)phenyl)methanol

A523067-1G A523067-5G
76790-102EA 23.34 USD
76790-102 76790-104
3-(Trifluoromethyl)benzhydrol 97%
3-(Trifluoromethyl)benzhydrol

Precaución: for Research Use Only

Formula: C₁₄H₁₁F₃O
MW: 252.23 g/mol
MDL Number: MFCD00014397
CAS Number: 728-80-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR