2-((tert-Butoxycarbonyl)amino)ethyl 4-methylbenzenesulfonate 97%

Supplier: Ambeed
Warning

A520891-1G A520891-5G A520891-25G
76790-080EA 12 USD
76790-080 76790-082 77669-302
2-((tert-Butoxycarbonyl)amino)ethyl 4-methylbenzenesulfonate 97%
2-((tert-Butoxycarbonyl)amino)ethyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₁₄H₂₁NO₅S
MDL Number: MFCD07776965
CAS Number: 158690-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR