Methyl-5-bromo-6-hydroxynicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 5-bromo-6-hydroxynicotinate

A517809-10G A517809-1G A517809-250MG A517809-25G A517809-5G
76790-060EA 246.13 USD
76790-060 76790-062 76790-064 76790-066 76790-068
Methyl-5-bromo-6-hydroxynicotinate 97%
Methyl-5-bromo-6-hydroxynicotinate

Precaución: for Research Use Only

Formula: C₇H₆BrNO₃
MW: 232.03 g/mol
MDL Number: MFCD08235247
CAS Number: 381247-99-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR