Trimethoxy(oct-7-en-1-yl)silane 95% stabilized

Supplier: Ambeed
Warning


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

A516116-1G A516116-5G
76790-048EA 23.34 USD
76790-048 76790-050
Trimethoxy(oct-7-en-1-yl)silane 95% stabilized
Trimethoxy(oct-7-en-1-yl)silane

Precaución: for Research Use Only

Formula: C₁₁H₂₄O₃Si
MW: 232.39 g/mol
MDL Number: MFCD00040026
CAS Number: 52217-57-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR