tert-Butyl propionate 98%

Supplier: Ambeed
Danger

Synonyms: Propionic Acid tert-Butyl Ester

A514525-25G A514525-5G
76790-040EA 91.24 USD
76790-040 76790-042
tert-Butyl propionate 98%
tert-Butyl propionate

Precaución: for Research Use Only

Formula: C₇H₁₄O₂
MW: 130.19 g/mol
MDL Number: MFCD00009304
CAS Number: 20487-40-5
UN: 3272
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR