(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile 98%

Supplier: Ambeed
Warning

Synonyms: 2-(1-Hydroxycyclohexyl)-2-(4-methoxyphenyl)acetonitrile, 1-[Cyano(4-methoxyphenyl)methyl]cyclohexanol

A514142-100MG A514142-1G A514142-250MG A514142-5G
76790-030EA 24 USD
76790-030 76790-032 76790-034 76790-036
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile 98%
(1-Hydroxycyclohexyl)(4-methoxyphenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₁₅H₁₉NO₂
MDL Number: MFCD06658142
CAS Number: 93413-76-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR