4-Bromo-2-(trifluoromethyl)quinoline 98%

Supplier: Ambeed
Warning

A508823-1G A508823-250MG A508823-5G
76790-000EA 54.11 USD
76790-000 76790-002 76790-004
4-Bromo-2-(trifluoromethyl)quinoline 98%
4-Bromo-2-(trifluoromethyl)quinoline

Precaución: for Research Use Only

Formula: C₁₀H₅BrF₃N
MW: 276.0526 g/mol
MDL Number: MFCD00277488
CAS Number: 18706-25-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR