trans-2-Dodecenoic acid 98%

Supplier: Ambeed
Danger

Synonyms: (E)-Dodec-2-enoic acid

A507643-250MG A507643-5G
76789-986EA 57.29 USD
76789-986 76789-988
trans-2-Dodecenoic acid 98%
trans-2-Dodecenoic acid

Precaución: for Research Use Only

Formula: C₁₂H₂₂O₂
MW: 198.31 g/mol
MDL Number: MFCD00045906
CAS Number: 32466-54-9
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR