3-Butenoic acid 95+% stabilized

Supplier: Ambeed
Danger

Synonyms: Vinylacetic Acid, 3-Butenoic Acid

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A507494-10G A507494-1G A507494-25G A507494-5G A507494-100G
76789-978EA 27.59 USD
76789-978 76789-980 76789-982 76789-984 77669-270
3-Butenoic acid 95+% stabilized
3-Butenoic acid

Precaución: for Research Use Only

Formula: C₄H₆O₂
MW: 86.09 g/mol
MDL Number: MFCD00002782
CAS Number: 625-38-7
UN: 3265
ADR: 8,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR