4-Chloro-3-(piperidine-1-carbonyl)phenylboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (4-Chloro-3-(piperidine-1-carbonyl)phenyl)boronic acid

A506950-1G A506950-250MG A506950-5G
76789-972EA 80 USD
76789-972 76789-974 76789-976
4-Chloro-3-(piperidine-1-carbonyl)phenylboronic acid 97%
4-Chloro-3-(piperidine-1-carbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₅BClNO₃
MW: 267.5200 g/mol
MDL Number: MFCD07783862
CAS Number: 871332-70-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR