5-Bromo-2-methyl-2H-indazole 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methyl-5-bromo-2H-indazole

A504487-10G A504487-1G A504487-250MG A504487-25G A504487-5G
76789-956EA 5.31 USD
76789-956 76789-958 76789-960 76789-962 76789-964 76789-966
5-Bromo-2-methyl-2H-indazole 97%
5-Bromo-2-methyl-2H-indazole

Precaución: for Research Use Only

Formula: C₈H₇BrN₂
MDL Number: MFCD09870047
CAS Number: 465529-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR