N-Methyl 3-boronobenzenesulfonamide 98%

Supplier: Ambeed
Warning

Synonyms: 3-(Methylsulphamoyl)phenylboronic acid, 3-Methylsulphamoylbenzeneboronic acid

A502271-100MG A502271-1G A502271-250MG A502271-5G
76789-944EA 37.14 USD
76789-944 76789-946 76789-948 77669-254
N-Methyl 3-boronobenzenesulfonamide 98%
N-Methyl-3-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₇H₁₀BNO₄S
MW: 215.0346 g/mol
MDL Number: MFCD07363845
CAS Number: 871329-75-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR