trans,trans-2,4-Decadienal 91%

Supplier: Ambeed
Warning

A500828-10G A500828-100G A500828-25G
76789-940EA 30 USD
76789-940 76789-942 77669-248 77872-837
trans,trans-2,4-Decadienal 91%
trans,trans-2,4-Decadienal

Precaución: for Research Use Only

Formula: C₁₀H₁₆O
MDL Number: MFCD00007007
CAS Number: 25152-84-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR