Ethyl-4-chloro-6-methoxyquinoline-3-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate, 4-Chloro-6-methoxyquinoline-3-carboxylic acid ethyl ester

A500713-100MG A500713-1G A500713-250MG A500713-5G
76789-932EA 53.05 USD
76789-932 76789-934 76789-936 76789-938
Ethyl-4-chloro-6-methoxyquinoline-3-carboxylate 95%
Ethyl 4-chloro-6-methoxyquinoline-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₂ClNO₃
MW: 265.70 g/mol
MDL Number: MFCD00173417
CAS Number: 22931-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR