4-(4-Bromopyrimidin-2-yl)morpholine 97%

Supplier: Ambeed
Warning

A500324-250MG A500324-5G
76789-914EA 31.83 USD
76789-914 76789-916
4-(4-Bromopyrimidin-2-yl)morpholine 97%
4-(4-Bromopyrimidin-2-yl)morpholine

Precaución: for Research Use Only

Formula: C₈H₁₀BrN₃O
MDL Number: MFCD03861153
CAS Number: 663194-10-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR